Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CIZ1 Polyclonal Antibody, Bethyl Laboratories

Catalog No. p-7140914
Encompass_Preferred
Change view
Click to view available options
Quantity:
20 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-156-0543 20 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-156-0543 Supplier Bethyl Laboratories, Inc Supplier No. A301497AT
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

The recommended shelf life for this product is 1 year from date of receipt.

May regulate the subcellular localization of CIP/WAF1.

Specifications

Antigen CIZ1
Applications Western Blot
Classification Polyclonal
Conjugate Unconjugated
Formulation TBS with 0.1% BSA and 0.09% sodium azide
Gene CIZ1
Gene Accession No. Q9ULV3
Gene Alias LSFR1, NP94, ZNF356, CDKN1A-interacting zinc finger protein 1, cip1-interacting zinc finger protein, nuclear protein NP94, zinc finger protein 356
Gene Symbols CIZ1
Host Species Rabbit
Immunogen Between 848 and 898
Purification Method Antigen affinity chromatography
Quantity 20 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 25792
Target Species Human
Content And Storage 4° C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.