Learn More
Apexbio Technology LLC CK-636, 10mg. Cas: 442632-72-6 MFCD: MFCD03036245. Arp2/3 complex inhibitor
SDP
Supplier:  Apexbio Technology LLC B305810
Encompass_Preferred
CK-636 is an Arp2/3 complex inhibitor with IC50 of 4 M, 24 M and 32 M for inhibition of actin polymerization induced by human, fission yeast and bovine Arp2/3 complex, respectively [1]. Other sizes are available. Please inqury us for quote.
Catalog No. 50-190-5342
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.