Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 alpha Recombinant Superclonal™ Antibody (5HCLC)

Catalog No. 711706
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-170-6 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-170-6 Supplier Invitrogen™ Supplier No. 711706
Add to Cart
Edge
Add to Cart

Rabbit Recombinant Superclonal Antibody

This antibody is predicted to react with Monkey, Rat, Pig. Recombinant rabbit Superclonal™ antibodies are unique offerings from Thermo Fisher Scientific. They are comprised of a selection of multiple different recombinant monoclonal antibodies, providing the best of both worlds - the sensitivity of polyclonal antibodies with the specificity of monoclonal antibodies - all delivered with the consistency only found in a recombinant antibody. While functionally the same as a polyclonal antibody - recognizing multiple epitope sites on the target and producing higher detection sensitivity for low abundance targets - a recombinant rabbit Superclonal™ antibody has a known mixture of light and heavy chains. The exact population can be produced in every lot, circumventing the biological variability typically associated with polyclonal antibody production. Note: Formerly called 'Recombinant polyclonal antibody', this product is now rebranded as 'Recombinant Superclonal™ antibody'. The physical product and the performance remain unchanged.

CNSK1A1 (CK1 alpha 1) is a member of the casein kinase I (CKI) gene family, which encodes serine/threonine kinases that preferentially phosphorylate acidic substrates. CKI proteins are monomeric, ranging from 25 to 55 kD, and are found in the nuclei, cytoplasm, and membrane fractions of eukaryotic cells. It can phosphorylate a large number of proteins. Participates in Wnt signaling. Phosphorylates CTNNB1 at 'Ser-45'. May phosphorylate PER1 and PER2. May play a role in segregating chromosomes during mitosis.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 alpha
Applications Immunocytochemistry
Classification Recombinant Superclonal
Clone 5HCLC
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.09% sodium azide; pH 7.2-7.4
Gene CSNK1A1
Gene Accession No. P48729
Gene Alias 2610208K14Rik; 4632404G05Rik; 5430427P18Rik; casein kinase 1 alpha 1; casein kinase 1 alpha 1 like; casein kinase 1 alpha 1 S homeolog; casein kinase 1 alpha 1 S homeolog; casein kinase 1, alpha 1; casein kinase 1 alpha isoform; casein kinase 1, alpha 1; casein kinase 1alpha S; casein kinase 1-alphaL; casein kinase 1-alphaLS; casein kinase I; casein kinase I alpha; casein kinase I alpha L isoform; casein kinase I alpha LS; casein kinase I alpha S; Casein kinase I isoform alpha; casein kinase I isoform alpha; casein kinase I; casein kinase I-alpha; CHUNP6894; CK I alpha; CK1; ck1 alpha; CK1a; ck1alpha; CK1alphaS; CKI alpha; CK-I alpha; CKIa; CKI-alpha; clock regulator kinase; Csnk1a; csnk1a1; CSNK1A1 protein; csnk1a1.S; CSNK1A1/CK1; CSNK1A1L; down-regulated in lung cancer; epididymis secretory sperm binding protein Li 77p; HEL-S-77p; HLCDGP1; OTTHUMP00000224000; OTTHUMP00000224001; OTTHUMP00000224003; PRO2975; protein kinase; sb:eu866; wu:fb65a02; wu:fi30h04; wu:fj19c11; XELAEV_18021437mg; YIL035C; zgc:92158
Gene Symbols CSNK1A1
Host Species Rabbit
Immunogen Protein corresponding to human CK1 alpha [aa2-aa337].
Purification Method Protein A
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1452
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.