Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CK1 delta Polyclonal Antibody
GREENER_CHOICE

Catalog No. PA5148874
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PA5148874 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PA5148874 Supplier Invitrogen™ Supplier No. PA5148874
  • $447.00
    $335.65 / Each of 1

    Save $111.35

    25% Off The following deviations and disclosures may produce prices below stated discounts but will not trigger price reductions: Price shown reflects a temporary, promotional price reduction. No other purchase or commercial obligation required to receive the reduced price. Price may change at any time. Promotional price valid on web orders only. Your contract pricing may differ. Other exclusions apply.
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Sheep Polyclonal Antibody

Reconstitute at 0.2 mg/mL in sterile PBS.

CSNK1D (CK1 delta), a serine/threonine kinase, plays a critical role in Wnt/beta-catenin signaling pathway. This pathway is important in both development and cancer. The encoded protein may also be involved in the regulation of apoptosis, circadian rhythm, microtubule dynamics, chromosome segregation, and p53-mediated effects on growth. The encoded protein is highly similar to the mouse and rat CK1 delta homologs. Three transcript variants encoding different isoforms have been found for CK1 delta.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CK1 delta
Applications Immunocytochemistry, Western Blot
Classification Polyclonal
Concentration 0.2 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and no preservative
Gene CSNK1D
Gene Accession No. P48730, Q06486, Q9DC28
Gene Alias 1200006A05Rik; AA409348; ASPS; casein kinase 1; casein kinase 1 delta; casein kinase 1, delta; casein kinase I isoform delta; CK1d; CK1delta; CKId; CKIdelta; CKI-delta; CSNK1D; D930010H05Rik; FASPS2; HCKID; HGNC:2452; Tau-protein kinase CSNK1D
Gene Symbols CSNK1D
Host Species Sheep
Immunogen E. coli-derived recombinant human CK1 delta , Arg316-Arg415.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 104318, 1453, 64462
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.