Learn More
Newcomer Supply CK5/14 + p63 + P504S (PIN-4) Control Slides, 10/set
SDP
Supplier:  Newcomer Supply 3651A
Encompass
Newcomer Supply CK5/14 + p63 + P504S Control Slides provide a useful combination of markers helpful in diagnosing prostatic intraepithelial neoplasia (PIN). For ease of screening, CK5/14 + p63 + P504S positive staining is in a single piece of prostate tissue. CK5/14 + p63 + P504S Control Slides contain sections of positive staining prostate and negative staining myometrium.
Catalog No. NC2101950
Only null left
Add to Cart
This item is not returnable. View return policy
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.