Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CKS1 Polyclonal Antibody
GREENER_CHOICE

Catalog No. 366800
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-680-0 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-680-0 Supplier Invitrogen™ Supplier No. 366800
Add to Cart
Edge
Add to Cart

Rabbit Polyclonal Antibody

CKS1 Polyclonal Antibody for Western Blot, IHC, ELISA, IP

CKS1B protein binds to the catalytic subunit of the cyclin dependent kinases and is essential for their biological function. The CKS1B mRNA is found to be expressed in different patterns through the cell cycle in HeLa cells, which reflects a specialized role for the encoded protein. At least two transcript variants have been identified for this gene, and it appears that only one of them encodes a protein.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CKS1
Applications ELISA, Immunohistochemistry, Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.25 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% sodium azide; pH 7.4
Gene CKS1B
Gene Accession No. P61024
Gene Alias 2410005G18Rik; 2610005D03Rik; AA407784; CDC28 protein kinase 1; CDC28 protein kinase 1B; CDC28 protein kinase regulatory subunit 1; CDC28 protein kinase regulatory subunit 1B; CDC2-associated protein CKS1; cell division control protein CKS1; CKS1; CKS-1; Cks1b; ckshs1; cyclin-dependent kinase regulatory subunit 1; cyclin-dependent kinases regulatory subunit 1; NB4 apoptosis/differentiation related protein; PNAS-143; PNAS-16; PNAS-18; RGD1561797; Sid 1334; sid1334; Unknown (protein for MGC:142665); zgc:101026
Gene Symbols CKS1B
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminal region of human and mouse Cks1.
Purification Method Antigen affinity chromatography
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1163
Target Species Human
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
WARNING: Reproductive Harm - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.