Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CL 075

Catalog No. 61425
Change view
Click to view available options
5mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No.
61-425 5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. 61-425 Supplier R&D Systems Supplier No. 6142/5
This item is not returnable. View return policy

TLR8 agonist

CL 075 is a TLR8 agonist. Induces production of TNF-α and IL-12 from peripheral blood mononuclear cells (PBMCs). Promotes maturation of monocyte-derived dendritic cells in combination with Poly(I:C) (Cat. No. 4287).

Specifications

Chemical Name or Material 2-Propylthiazolo[4,5-c]quinolin-4-amine
CAS 256922-53-9
Assay Percent Range >98%
Quantity 5 mg
Molecular Formula C13H13N3S
Formula Weight Observed MW: 243.33
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.