Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Thermo Scientific™ CL-XPosure™ Film
GSA_VA

Catalog No. p-200000282
Change view
Click to view available options
Dimensions (LxW):
18 x 13 cm
24 x 18 cm
25 x 20 cm
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Dimensions (LxW)
PI34090 18 x 13 cm
PI34089 24 x 18 cm
PI34091 25 x 20 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. PI34090 Supplier Thermo Scientific™ Supplier No. 34090
Only null left
Add to Cart
Add to Cart

Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based western blots and other chemiluminescent protein or nucleic acid assay methods.

Thermo Scientific CL-XPosure Film is an economically priced clear-blue X-ray film especially for detection and documentation of ECL-based western blots and other chemiluminescent protein or nucleic acid assay methods. CL-XPosure Film is an excellent photographic film for use with enhanced chemiluminescence (ECL) substrates for horseradish peroxidase (HRP) or alkaline phosphatase (AP). Convenient sizes are offered for protein western blotting, standard film cassettes, and film developing equipment. This autoradiography film also works well for chemiluminescent or isotopic DNA and RNA methods, including Southern and northern blotting and gel-shift assays (EMSA). The quality and price of this film make it the ideal choice for life science research applications.

Features include:
• High performance—just as sensitive as other commercially available autoradiography films
• Economical—achieve high-quality images at an affordable price
• Convenient—available in five sheet sizes in packages containing 100 non-interleaved sheets
• Perfect match—validated for use with SuperSignal chemiluminescent substrates and Pierce ECL and ECL Plus chemiluminescent substrates

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Product Type X-Ray Film
Length (English) 7 in.
Width (English) 5 in.
Quantity 100 Sheets
Content And Storage Store at room temperature.
Dimensions (LxW) 18 x 13 cm
Product Line CL-Xposure
WARNING: Cancer - www.P65Warnings.ca.gov
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.