Learn More
ATCC Cladosporium sp. (ATCC 8462) - Frozen ampoule, research culture (NCTC 4526)
Supplier: ATCC 8462
ATCC 8462 is a frozen research culture of Cladosporium sp. supplied as a frozen ampoule containing viable cells suspended in cryoprotectant. The product is intended for laboratory research use only and requires appropriate biosafety practices for handling.
- Frozen ampoule format containing viable fungal cells in cryoprotectant.
- Store at -80°C or colder; vapor phase liquid nitrogen preferred for long-term storage.
- Recommended recovery medium: potato carrot agar (ATCC Medium 335).
- Incubate at 25°C and inspect for growth for up to 4 weeks.
- Biosafety level 1; use appropriate PPE and institutional biosafety procedures.