Learn More
Malvern Panalytical CLAISSE HEAVY MOLD 32MM 36G
Supplier:  Malvern Panalytical P023201
Encompass
CLAISSE HEAVY MOLD, 32 MM, 36G , 1,2 MM THICKHeavy Mold, 32 mm, 36g Pt-Au(5%) mold, having an internal diameter of 32 mm,a nominal weigh (± 5%) of 36 g and an internal height of 4,8 mm. Used for glassdisk production. This platinum-gold alloy is universally accepted as the material ofchoice for crucibles and casting molds for XRF analysis. It has higher temperatureresistance than pure platinum and has a non-wetting property that allows an easyremoval of sample after fusion.
Catalog No. NC2391965
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.