Learn More
Eppendorf™ Clamp Bundle for Innova 42R
Catalog No. M11909921 Shop All Eppendorf North America Products
Change view
Click to view available options
For Use With (Equipment):
Innova™ 42R
Catalog No. For Use With (Equipment)
M11909921 Innova™ 42R
1 options
Catalog No. M11909921 Supplier Eppendorf™ Supplier No. M11909921
Add to Cart
Add to Cart
This Clamp Bundle for Innova 42R features flask clamps that secure a wide range of vessels to the shaker platform and are constructed of a single piece stainless-steel for added strength and safety. Flask clamps come with required screws and are fitted with coiled wire springs for added stability.
Pre-selected clamps for the Innova 42R Universal Platform includes:
  • 4 x 125 mL clamp
  • 4 x 250 mL clamp
  • 4 x 500 mL clamp
  • 4 x 1000 mL clamp
TRUSTED_SUSTAINABILITY
Specifications
Holds 125/250/500/1000 mL
Product Type Universal Platform Clamps
Material Stainless Steel
For Use With (Equipment) Innova™ 42R
Packaging Quantity 16 Pk.