Learn More
Thermo Scientific™ Clamps for CO2 Resistant Shaker
Achieve maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
$30.15 - $633.50
Catalog Number | Mfr. No. | Product Type | Clamps | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Clamps | Price | Quantity & Availability | ||||
88-881-105
|
Thermo Scientific™
88881105 |
Flask Clamp | 50 mL |
Each for $30.15
|
|
||||
88-881-106
|
Thermo Scientific™
88881106 |
Flask Clamp | 100 mL |
Each for $34.45
|
|
||||
88-881-107
|
Thermo Scientific™
88881107 |
Flask Clamp | 150 mL |
Each for $41.55
|
|
||||
88-881-108
|
Thermo Scientific™
88881108 |
Flask Clamp | 200 mL |
Each for $45.90
|
|
||||
88-881-109
|
Thermo Scientific™
88881109 |
Flask Clamp | 250 mL |
Each for $49.75
|
|
||||
88-881-110
|
Thermo Scientific™
88881110 |
Flask Clamp | 500 mL |
Each for $74.40
|
|
||||
88-881-111
|
Thermo Scientific™
88881111 |
Flask Clamp | 1000 mL |
Each for $125.50
|
|
||||
88-881-112
|
Thermo Scientific™
88881112 |
Flask Clamp | 2000 mL |
Each for $185.30
|
|
||||
88-881-113
|
Thermo Scientific™
88881113 |
Flask Clamp | 3000 mL |
Each for $221.50
|
|
||||
88-881-114
|
Thermo Scientific™
88881114 |
Flask Clamp | 5000 mL |
Each for $305.50
|
|
||||
88-881-115
|
Thermo Scientific™
88881115 |
Infusion bottle clamp | 500 mL |
Each for $76.80
|
|
||||
88-881-116
|
Thermo Scientific™
88881116 |
Infusion bottle clamp | 1000 mL |
Each for $122.80
|
|
||||
88-881-117
|
Thermo Scientific™
88881117 |
Funnel Clamp | Separatory |
Each for $519.50
|
|
||||
88-881-133
|
Thermo Scientific™
88881133 |
Starter Kit | Flask |
Each for $633.50
|
|
||||
Description
Maximize capacity on the Thermo Scientific™ CO2 Resistant Shaker universal platform with a variety of clamps to fit most vessels.
- Compatible with Thermo Scientific™ CO2Resistant Shakers, IntelliStack™ Incubator Shakers, and IntelliStack™ CO2 Incubator Shakers.
- Refer to the user manual or brochure for details on the maxinum number of clamps that can fit per platform.