Learn More
Description
Includes
Single Tray only
- Multiplex up to 8 samples
- No vacuum source needed
- No power supply needed
- Yellow
Specifications
Specifications
| Kit Contents | Single Tray Only |
| Content And Storage | Room Temperature |
| Particle Size | Not Applicable |
| Quantity | 1 Ea. |
| For Use With (Application) | HPLC |
| Type | Tools and Accessories |
| Sufficient For | Analytical Sample Preparation, Clarification |
| Format | 8-well Strips |
| Capacity (Metric) | 600 μL |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.