Learn More
Description
Includes
NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet
Inorganic Standard manufactured for AA, ICP, ICP/MS, IC, XRF, and other analytical instrumentation
| Analyte | Concentration |
|---|---|
| Cu 65 | 10μg/mL |
Certifications
Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001
Compliance
GHS/OSHA Complaint
Specifications
Specifications
| Physical Form | Liquid |
| CAS Max % | 2.0, 98.0, 0.0 |
| Quantity | 125 mL |
| Packaging | Triple-leached LDPE bottle |
| Grade | Claritas PPT |
| Concentration or Composition (by Analyte or Components) | 10mg/L |
| Chemical Name or Material | Copper 65 Isotope Standard, For ICP/MS |
Safety and Handling
| DOTInformation | Hazardous |
| Recommended Storage | Ambient |
Product intended for laboratory use only
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
