Learn More
Abcam Clarithromycin, Macrolide Antibiotic, Solid, 98% Purity, 100 mg
Description
- Molecular Weight: 748 Da
- Purity: >98%
- Form: Solid
- Mechanism: Interferes with protein synthesis by reversibly binding to the 50S subunit in the bacterial ribosome
- Biological Activity: Regulates matrix metalloprotease activity; exhibits bacteriostatic and bactericidal effects in vitro and in vivo
The product is subject to the following: Abcam Restricted Use Statement
Specifications
Specifications
| Shipping Condition | Ambient |
| Target Molecule | 50S Ribosomal Subunit, Matrix Metalloprotease |
| Content And Storage | Store at 4°C. |
| Form | Solid |
| Formulation | 100% Clarithromycin |
| Quantity | 100 mg |
| For Use With (Application) | Protein synthesis inhibition, Matrix metalloprotease regulation |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.