Learn More
TOKU-E CLARITHROMYCIN SOLN 1ML
Supplier:  TOKU-E C2881ML
Encompass_Preferred
Antibiotic, ReadyMade™, sterile-filtered, 1 mg/ml in water. Useful in cancer research, cell culture, susceptibility testing.
Catalog No. NC2068756
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.