Learn More
R&D Systems™ ClariTSA Fluorophore Kit 620

Description
ClariTSA™ reagents are exceptionally bright dyes that offer an effective way to boost signal intensity and detect low-abundance targets in spatial biology applications. They are designed for use in ISH, IHC and ICC experiments where in situ detection of target protein or nucleic acid sequences is required. ClariTSA reagents have been specifically optimized to deliver exceptional signal-to-noise in the RNAscope™ Multiplex Fluorescent v2 Assay for visualization of gene expression at the single cell level. They are suitable for multiplexing and can be combined with DAPI (Cat. No. 5748) counter-staining.
The basis for signal amplification comes from the specific and high-density deposition of ClariTSA dyes adjacent to a horseradish peroxidase (HRP)-labeled probe in a process termed Tyramide Signal Amplification (TSA) or Catalyzed Reporter Deposition (CARD).
Excitation maximum = 591 nm; emission maximum = 615 nm
Please see the product datasheet or protocol for information on product use.
ClariTSA™ is a trademark of Bio-Techne Corp.
Specifications
Specifications
| Content And Storage | Store at -20°C |
| Product Line | Tocris |
| Quantity | 1 Kit (20-80 Slides) |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.