Learn More
Diagnostic Test Group CLARITY MONO CASSETTE BX15
SDP
Supplier:  Diagnostic Test Group DTGMONO
Encompass_Preferred
Sensitivity: >= 99%; Specificity: 98.6%; 99% Overall Accuracy; Fast and Easy Test Results in 8 Minutes; No Age Restriction; 18 Month Dating; CLIA Waived for Whole Blood Only, can be used with Plasma/Serum; Clarity Monoucleousis Kit (DTG-MONO) Contains: 15-Test Cassettes, 1-Developer Solution, 15-Sample Transfer Tubes, 1-Positive Control, 1-Negative Control, Package Insert, and Procedure Card.
Catalog No. 50-143-058
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.