Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

CLASP1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
SDP

Catalog No. 501733095 Shop All Proteintech Group Inc Products
Encompass_Preferred
Change view
Click to view available options
Quantity:
150 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-173-3095 150 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-173-3095 Supplier Proteintech Group Inc Supplier No. 208031AP150UL
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

CLASPs, such as CLASP1, are nonmotor microtubule-associated proteins that interact with CLIPs.

Specifications

Antigen CLASP1
Applications Immunocytochemistry, Immunofluorescence, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot
Classification Polyclonal
Concentration 0.23 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CLASP1
Gene Accession No. Q7Z460, Q80TV8
Gene Alias CLASP1, CLIP associating protein 1, hOrbit1, KIAA0622, MAST1, Multiple asters homolog 1, Protein Orbit homolog 1
Gene Symbols Clasp1
Host Species Rabbit
Immunogen CLASP1 Fusion Protein Ag14724
Purification Method Antigen Affinity Chromatography
Quantity 150 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 23332, 304740, 76707
Target Species Human, Mouse, Rat
Content And Storage -20°C
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.