Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

National Safety Apparel™ Enespro™ Class 0 Voltage 11 in. Glove Kit
SDP

Catalog No. 17355024 Shop All National Public Seating Corporation Products
Change view
Click to view available options
Color:
Black
Red
Yellow
Size:
8
9
10
11
12
15 product options available for selection
Product selection table with 15 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
17-355-024 Red 12
17-355-015 Black 8
17-355-016 Black 9
17-355-017 Black 10
17-355-018 Black 11
17-355-019 Black 12
17-355-020 Red 8
17-355-021 Red 9
17-355-022 Red 10
17-355-023 Red 11
17-355-025 Yellow 8
17-355-026 Yellow 9
17-355-027 Yellow 10
17-355-028 Yellow 11
17-355-029 Yellow 12
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 15 options available.
15 options
This item is not returnable. View return policy
Catalog No. 17-355-024 Supplier National Safety Apparel™ Supplier No. KITGC0R12
Add to Cart
Add to Cart
This item is not returnable. View return policy

Kit includes a pair rubber electrical insulating gloves, a pair of 10 in. leather glove protectors, and a convenient storage bag. Class 0 insulating gloves will protect workers from up to 1000 V.

  • Electrical insulating gloves protect against electrical shock and other arc flash hazards
  • Wear with included leather glove protectors for additional resistance against mechanical hazards
  • Rubber voltage gloves are electrically tested and marked with initial test date
  • Class 0 - Max Use Voltage 1,000V AC/1,500V DC

Specifications

Certifications/Compliance ANSI/ASTM D120, Class 0, Type I, NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F696
Applications Industrial
Color Red
Hand Specific
Material Rubber, Leather
Product Type Electrical Insulating Gloves Kit
Size 12
For Use With (Application) Utilities, Manufacturing, Electrical Contractors
Glove Cuff Style Straight
Includes Natural Rubber Electrical Insulating Gloves, Leather Glove Protectors, and Storage Bag
Series KITGC0R
Length (English) 11 in.
Length (Metric) 28 cm
Voltage 1000 V
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.