Learn More
National Safety Apparel™ Enespro™ Class 2 Voltage 14 in. Glove Kit
Shop All National Public Seating Corporation Products

Description
- Electrical insulating gloves protect against electrical shock and other arc flash hazards
- Wear with included leather glove protectors for additional resistance against mechanical hazards
- Rubber voltage gloves are electrically tested and marked with initial test date
- Class 2 - Max Use Voltage 17,000V AC/25,500V DC
- Gloves should be used within one year of marked test date
Specifications
Specifications
| Certifications/Compliance | ANSI/ASTM D120, Class 2, Type I, NFPA 70E, CSA Z462, OSHA 1910.269, ASTM F696 |
| Applications | Industrial |
| Color | Yellow |
| Hand | Specific |
| Material | Rubber, Leather |
| Product Type | Electrical Insulating Gloves Kit |
| Size | 11 |
| For Use With (Application) | Utilities, Manufacturing, Electrical Contractors |
| Glove Cuff Style | Straight |
| Includes | Class 2 Electrical Insulating Gloves, Leather Glove Protectors, and a Storage Bag |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.