Learn More
Honeywell™ Class 2 Traffic Safety Vests
Catalog No. 19120617
Change view
Click to view available options
Color:
Lime
Orange
Size:
2X-Large
3X-Large
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Size
19-120-617 Orange 2X-Large
19-120-620 Lime 3X-Large
19-120-619 Lime 2X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Catalog No. 19-120-617 Supplier Honeywell™ Supplier No. TVA3/2NZ/O2XL
This item is not returnable. View return policy
Provides excellent visibility for workers exposed to traffic speeds exceeding 50 mph.
  • Constructed of 100% polyester
  • Has two, 1 1/2 in. (3.8cm) North-Brite™ reflective vertical stripes on the front and back
  • With a pair of 1 1/2 in. (3.8cm) horizontal stripes on the waist
  • Zipper front makes donning quick and easy
Order Info
Available in orange or lime.
Compliance
Meets ANSI/ISEA 107, Class 2 standards.
TRUSTED_SUSTAINABILITY
Specifications
Color Orange
Size 2X-Large
Material Polyester