Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific Supplies Class 3R Laser Pointers
SDPSCI_ED

Catalog No. S27811 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Description:
Green
Red
Red/Green
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Description
S27811 Green
S27840 Red
S27797 Red/Green
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. S27811 Supplier United Scientific Supplies Supplier No. GLP006
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Ideal for optical experiments

  • Class 3R Laser Pointer with push-button switch
  • Ideal for optical experiments
  • Housed in rubberized aluminum
  • Pen-style case with pocket clip
  • Dimensions (L x D): 6 x 0.5 in.
  • Maximum Output: 5 mW
  • Wavelength: 532 nm
  • Operates on (2) AAA batteries
  • Available in Red, Green, Red/Green, and Violet

Specifications

Product Type Laser Pointer, Class 3R
Description Green
For Use With (Application) Optical Experiments
Length (English) 6 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.