Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

United Scientific™ Class A Automatic Burette Bottle
SDPSCI_ED

Catalog No. S00764 Shop All United Scientific Supplies, Inc Products
Change view
Click to view available options
Capacity (Metric):
2000 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Capacity (Metric)
S00764 2000 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. S00764 Supplier United Scientific™ Supplier No. BRB2000
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Burette bottle for use with Class A Automatic Burette.

  • Compatible with the 25 mL and 50 mL automatic burettes

Specifications

Color Clear
Capacity (Metric) 2000 mL
Length (Metric) 263 mm
Material Glass
Includes Glass bottle with 29/32 ground glass joint
Type Buret
Class Class B
Certifications/Compliance ASTM E287, Individually Serialized and Certified
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.