Learn More
Labconco™ Class II Biosafety Cabinet IV Bar Kit
IV bar, hooks and hardware to support intravenous solution bottles and bags in Logic Biosafety Cabinets.
Supplier: Labconco™ 3288901
Description
Kit includes:
- IV bar
- Mounting hardware
- Four hangers (2 short, 2 long)
- For use with Optic White Labconco Class II Biosafety Cabinets
- Compatible with Legacy Purifier Logic biosafety cabinets
- Installation is required
Specifications
| 122.4 cm | |
| 0.75 in. | |
| 122.4 cm | |
| 1.9 cm | |
| 4 ft. Optic White Labconco Logic Biosafety Cabinets | |
| 1.9 cm |
| IV Bag Holder Kit | |
| 48.2 in. | |
| IV Bar, 2 Short Hooks, 2 Long Hooks, Installation Hardware | |
| 48.2 in. | |
| 0.75 in. |