Description
- Wide profile and brim increase protection from sun and improve ventilation
- Constructed of high-density polyethylene
- Accessory slots accept most attachments, including welding helmets, earmuffs, faceshields, and communication devices
- Six-point suspension is either a standard design with sizing posts or a Flex-Gear™ ratchet design with sizing knob
- Generous cotton browpad is absorbent and fast drying; brow pad is replaceable
Compliance
Meet ANSI™/ISEA Z89.1-2003 Type I, Class G and E requirements.
Specifications
Specifications
| Certifications/Compliance | Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C |
| Color | Hi-vis Green |
| Material | HDPE |
| Suspension Type | 6-Point Pin-Lock |
Safety Services
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.