Learn More
Bullard™ Classic Series C30 Hardhats
SDP
Catalog No. 19098003 Shop All Bullard Company Products
Change view
Click to view available options
Certifications/Compliance:
Meets ANSI Z89.1-2003; Type I; Class G and E.
Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C
Color:
Blue
Green
Hi-vis Green
Hi-vis Orange
Hi-vis Yellow
Orange
Tan
White
Yellow
Suspension Type:
6-Point Pin-Lock
6-Point Ratchet
6-Point Standard
13 product options available for selection
Product selection table with 13 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Certifications/Compliance Color Suspension Type
19-098-003 Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C Hi-vis Green 6-Point Ratchet
19-047192 Meets ANSI Z89.1-2003; Type I; Class G and E. Orange 6-Point Ratchet
19-085439 Meets ANSI Z89.1-2003; Type I; Class G and E. Green 6-Point Ratchet
19-021362 Meets ANSI Z89.1-2003; Type I; Class G and E. Yellow 6-Point Ratchet
19-021361 Meets ANSI Z89.1-2003; Type I; Class G and E. White 6-Point Ratchet
19-098-005 Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C Hi-vis Yellow 6-Point Ratchet
19-021350 Meets ANSI Z89.1-2003; Type I; Class G and E. Blue 6-Point Standard
19-098-001 Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C Hi-vis Orange 6-Point Pin-Lock
19-098-002 Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C Hi-vis Yellow 6-Point Pin-Lock
19-021353 Meets ANSI Z89.1-2003; Type I; Class G and E. Orange 6-Point Standard
19-098-000 Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C Hi-vis Green 6-Point Pin-Lock
19-021356 Meets ANSI Z89.1-2003; Type I; Class G and E. Yellow 6-Point Standard
19-021357 Meets ANSI Z89.1-2003; Type I; Class G and E. Tan 6-Point Pin-Lock
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 13 options available.
13 options
Catalog No. 19-098-003 Supplier Bullard™ Supplier No. 30HGR
Add to Cart
Add to Cart
Offer a comfortable and secure fit
  • Wide profile and brim increase protection from sun and improve ventilation
  • Constructed of high-density polyethylene
  • Accessory slots accept most attachments, including welding helmets, earmuffs, faceshields, and communication devices
  • Six-point suspension is either a standard design with sizing posts or a Flex-Gear™ ratchet design with sizing knob
  • Generous cotton browpad is absorbent and fast drying; brow pad is replaceable
Compliance
Meet ANSI™/ISEA Z89.1-2003 Type I, Class G and E requirements.
Specifications
Certifications/Compliance Meets or exceeds ANSI Z891-1997; Type 1; Class E G and C
Color Hi-vis Green
Material HDPE
Suspension Type 6-Point Ratchet
Adhere to
Safety Regulations and Best Practices
Take advantage of a range of Safety Services from our trusted suppliers to help ensure the safety and compliance of your people, processes, products, and facilities. Services include assessments, training, equipment maintenance, safety programs, and more.