Learn More
Classroom Rock and Mineral Collection
SDPSCI_ED
Catalog No. S84011
Click to view available options
Quantity:
50 Specimens
Catalog No. Quantity
S84011 50 Specimens
1 options
Catalog No. S84011 Supplier Geoscience Industries Supplier No. 125000S
Includes
Manual and description key
50 rock and mineral specimens.
Samples have been carefully selected to support studies of:
  • Rock-forming minerals
  • Igneous, metamorphic, and sedimentary comparison
  • Ores of common metals
  • Sources of uranium
  • Hardness scale
Specimen dimensions (approx.): 1 x 1 in.
Specifications
Includes Manual and description key
Quantity 50 Specimens
Specimen Size 1 x 1 in.