Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Clathrin Heavy Chain Monoclonal Antibody (X22), Alexa Fluor™ 488
GREENER_CHOICE

Catalog No. PIMA1065A48 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
50 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIMA1065A48 50 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIMA1065A48 Supplier Invitrogen™ Supplier No. MA1065A488
Add to Cart
Edge
Add to Cart

Mouse Monoclonal Antibody

MA1065A488 detects clathrin heavy chain in non-human primate, bovine, human, rat and mouse tissues as well as hamster (CHO) cells. The MA1065A488 immunogen is purified human brain clathrin heavy chain. Electron microscopy and proteolysis mapping demonstrate that MA1065A488 binding occurs towards the central hub of the triskelion, N-terminal to the light chain binding regions.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Clathrin Heavy Chain
Applications Immunocytochemistry
Classification Monoclonal
Clone X22
Concentration 1 mg/mL
Conjugate Alexa Fluor 488
Formulation PBS with 1mg/mL BSA and 0.05% sodium azide; pH 7.4
Gene CLTCL1
Gene Accession No. P11442, P49951, P53675, Q00610, Q68FD5
Gene Alias 3110065L21Rik; CHC; CHC17; CHC22; clathrin heavy chain; clathrin heavy chain 1; Clathrin heavy chain 2; clathrin heavy chain like 1; clathrin heavy chain on chromosome 17; Clathrin heavy chain on chromosome 22; clathrin, heavy chain (Hc); clathrin, heavy chain-like 1; clathrin, heavy polypeptide (Hc); Clathrin, heavy polypeptide D; clathrin, heavy polypeptide-like 1; clathrin, heavy polypeptide-like 2; CLH17; CLH-17; CLH22; CLH-22; Cltc; CLTCL; CLTCL1; CLTCL2; CLTD; Hc; KIAA0034; R74732
Gene Symbols CLTC, CLTCL1
Host Species Mouse
Immunogen Purified human brain clathrin heavy chain.
Purification Method Protein A
Quantity 50 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100756644, 1213, 281080, 54241, 67300, 8218
Target Species Human, Mouse, Rat, Monkey, Bovine, Hamster
Content And Storage 4°C, store in dark, DO NOT FREEZE!
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.