Learn More
Invitrogen™ Claudin 1 Polyclonal Antibody

Description
This antibody is predicted to react with bovine, canine, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is skin, breast carcinoma or ovarian carcinoma.
Specifications
Specifications
| Antigen | Claudin 1 |
| Applications | Immunohistochemistry (Frozen), Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry |
| Classification | Polyclonal |
| Concentration | Conc. not determined |
| Conjugate | Unconjugated |
| Formulation | PBS with 1% BSA and 0.1% sodium azide; pH 7.6 |
| Gene | CLDN1 |
| Gene Accession No. | O88551, O95832 |
| Gene Alias | AI596271; claudin 1; claudin-1; CLD1; CLDN1; hypothetical protein; ILVASC; SEMP1; Senescence-associated epithelial membrane protein; senescence-associated epithelial membrane protein 1; tight junction protein; UNQ481/PRO944 |
| Gene Symbols | CLDN1 |
| Show More |
Safety and Handling
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.