Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 10 Polyclonal Antibody

Catalog No. p-7171848 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
500 μL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
01-673-307 1 mL
PIPA532351 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 01-673-307 Supplier Invitrogen™ Supplier No. PA137476
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, canine, and mouse based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is brain, skeletal muscle or ovarian carcinoma.

Claudin proteins are a family of proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different Claudin composition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 10
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Formulation PBS with 1% BSA and 0.1% sodium azide; pH 7.6
Gene Cldn10
Gene Accession No. P78369
Gene Alias 6720456I16Rik; claudin 10; claudin 10A; claudin 10B; claudin-10; claudin-10A; Cldn10; Cldn10a; Cldn10b; CPETRL3; D14Ertd728e; Oligodendrocyte-specific protein-like; OSP-L; OSP-like; OSP-like protein
Gene Symbols Cldn10
Host Species Rabbit
Immunogen Synthetic peptide from C-terminus of human claudin-10 protein.
Purification Method Antigen Affinity Chromatography
Quantity 1 mL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9071
Target Species Human
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.