Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 18.2 Recombinant Mouse Monoclonal Antibody (hu7V3)

Catalog No. MA554531
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA554531 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA554531 Supplier Invitrogen™ Supplier No. MA554531
Only null left
Add to Cart
Add to Cart

Mouse Recombinant Monoclonal Antibody

Compatible with Direct ELISA.

Plays a major role in tight junction-specific obliteration of the intercellular space, through calcium-independent cell-adhesion activity.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 18.2
Applications ELISA
Classification Recombinant Monoclonal
Clone hu7V3
Concentration 1.71 mg/mL
Conjugate Unconjugated
Formulation PBS with no preservative; pH 7.4
Gene CLDN18
Gene Accession No. P56856
Gene Alias SFTA5; SFTPJ
Gene Symbols CLDN18
Host Species Mouse
Immunogen Recombinant Human CLDN18.2.
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 51208
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG2a mFc VHH
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.