Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Claudin 3 Polyclonal Antibody, Invitrogen™

Catalog No. PIPA532353
Change view
Click to view available options
Quantity:
500 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PIPA532353 500 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PIPA532353 Supplier Thermo Scientific Supplier No. PA532353

Rabbit Polyclonal Antibody

This antibody is predicted to react with bovine, canine, mouse, porcine and rat based on sequence homology. Heat-mediated antigen retrieval is recommended prior to staining, using a 10mM citrate buffer, pH 6.0, for 10 minutes followed by cooling at room temperature for 20 min. Following antigen retrieval, incubate samples with primary antibody for 10 min at room temperature. A suggested positive control is small intestine tissue.

Claudin 1-8 proteins are a family of transmembrane proteins associated with tight junctions. Tight junctions are specialized regions of cell to cell contact; made up of a network of strands to act as a molecular gasket for preventing the leakage of ions, water etc. between cells. They are abundant in luminal epithelial sheets where they maintain epithelial cell polarity. Different tissues exhibit different claudin composition.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 3
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Unconjugated
Gene CLDN3
Gene Accession No. Q63400, Q9Z0G9, O15551
Gene Alias C7orf1, CPE-R2, CPETR2, HRVP1, RVP1, CPE-R 2, CPE-receptor 2, Clostridium perfringens enterotoxin receptor 2, claudin-3, ventral prostate.1 protein homolog, ventral prostate.1-like protein
Gene Symbols CLDN3
Host Species Rabbit
Immunogen Synthetic peptide derived from the C-terminus of mouse Claudin-3
Purification Method Antigen affinity chromatography
Quantity 500 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 1365, 65130, 12739
Target Species Chicken, Human, Mouse, Pig, Rat
Content And Storage -20°C or -80°C if preferred
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.