Learn More
IBL America Claudin-6 (C)Anti-Mouse Rabbit IgG Affinity Purify
Supplier:  IBL America 18865
Encompass
For research use only, not for use in diagnostic procedures.This antibody can be stained in frozen tissues by several immunohistochemicaltechniques such as immunofluoroscent method. The optimal dilution is about5 ug/mL, however, the dilution rate should be optimized by each laboratories. This antibody can be used for western blotting in concentration of about 1ug/mL.
Catalog No. NC1822484
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.