Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ Claudin 8 Monoclonal Antibody (474210)
GREENER_CHOICE

Catalog No. MA561537
Encompass_Preferred
Change view
Click to view available options
Quantity:
100 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
MA561537 100 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. MA561537 Supplier Invitrogen™ Supplier No. MA561537
Only null left
Add to Cart
Add to Cart

Mouse Monoclonal Antibody

Reconstitute at 0.5 mg/mL in sterile PBS.

Claudin-8 belongs to the claudin family of integral membrane proteins. Claudins are responsible for the formation of tight junction strands, and are connected with the actin cytoskeleton mediated by ZO-1. Claudin-8 is a tight junction protein expressed in several distal nephron segments, suggesting that it may have a role as a paracellular cation barrier protein. Claudin-8 is expressed in the inner ear, and the umbrella cell tight junctions in the bladder epithelium.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Claudin 8
Applications Flow Cytometry
Classification Monoclonal
Clone 474210
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 5% trehalose and no preservative
Gene CLDN8
Gene Accession No. P56748
Gene Alias AI648025; claudin 8; claudin-8; CLDN8; epididymis secretory protein Li 79; HEL-S-79; UNQ779/PRO1573
Gene Symbols CLDN8
Host Species Mouse
Immunogen HEK293 human embryonic kidney cell line transfected with human Claudin-8, Met1-Val225 (Ser151Pro).
Purification Method Protein A/G
Quantity 100 μg
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 9073
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Lyophilized
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.