Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Invitrogen™ CLBT Polyclonal Antibody
GREENER_CHOICE

Catalog No. FEPA5144664
Change view
Click to view available options
Quantity:
100 μL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
FEPA5144664 100 μL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. FEPA5144664 Supplier Invitrogen™ Supplier No. PA5144664
Only null left
Add to Cart
Add to Cart

Rabbit Polyclonal Antibody

Positive control: MCF-7 cell lysate, Hela cell lysate, 293T cell lysate, human placenta tissue lysate, rat testis tissue, SiHa. Predicted band size: 25 kDa Subcellular Location: Cytoplasmic vesicle membrane, Membrane, coated pit.

Clathrin is the major protein of the polyhedral coat of coated pits and vesicles. Two different adapter protein complexes link the clathrin lattice either to the plasma membrane or to the trans-Golgi network.
TRUSTED_SUSTAINABILITY

Specifications

Antigen CLBT
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 1 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4
Gene CLTB
Gene Accession No. P08082, P09497
Gene Alias 2310046E19Rik; Clathrin light chain B; clathrin light chain B pseudogene; clathrin, light chain (Lcb); clathrin, light chain B; clathrin, light polypeptide (Lcb); Cltb; Lcb
Gene Symbols CLTB
Host Species Rabbit
Immunogen Recombinant protein within human CLTB aa 30-229/229.
Purification Method Affinity Chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 116561, 1212
Target Species Human, Rat
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.