Learn More
Description
- Clean Room
- Latex-Free
Compliance
Class 10 Certification
Specifications
Specifications
| Material | Latex-free |
| Color | Blue |
| Dimensions (L x W) | 8.5 x 11 in. |
| Plain paper | Yes |
| Weight (English) | 28# |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.