Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

NOTRAX™ Clean-Step™ Frame

Catalog No. 19085977 Shop All NoTrax Products
Change view
Click to view available options
Length (English):
20 in.
26 in.
28 in.
Width (English):
38 in.
47 in.
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Length (English) Width (English)
19-085-977 20 in. 38 in.
19-085-978 20 in. 47 in.
19-085-982 26 in. 38 in.
19-085-986 28 in. 47 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 19-085-977 Supplier NOTRAX™ Supplier No. 080S203800
Only null left
Add to Cart
Add to Cart

For use with Clean-Step™ Tacky Mat

Specifications

For Use With (Equipment) For Clean-Step™ Tacky Mat
Length (English) 20 in.
Quantity 1 Ea.
Width (English) 38 in.
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.