Learn More
3M™ Clean-Trace™ Surface ATP, Sold by AquaPhoenix Scientific
SCI_ED
Catalog No. S27250 Shop All AquaPhoenix Scientific Products
Change view
Click to view available options
No. per Case:
100/Cs.
Catalog No. No. per Case
S27250 100/Cs.
1 options
Catalog No. S27250 Supplier 3M™ Supplier No. UXL100
Add to Cart
Add to Cart
Tests for ATP to verify cleaning effectiveness
  • Provides a tool to help assess and improve hygienic performance and reduce risk
  • Single use test contains swab for collection of sample from surface
  • Pre-moistened to aid sample collection
  • Easy to use and simple to learn
Specifications
Includes Swab
For Use With (Application) Detecting ATP to verify cleanliness
No. per Case 100/Cs.
Product Type Clean-Trace™ Surface ATP