Learn More
Med Vet International Cleaner Disinfectant Cavicide Bottle, 12/Case
Supplier:  Med Vet International MAP131024CS
Encompass_Preferred
CaviCide is a convenient, ready-to-use, intermediate-level surface disinfectant which is effective against TB, HBV, HCV, viruses (hydrophilic and lipophilic), bacteria (including MRSA and VRE) and fungi. It is safe for all areas of the facility including NICU, operating rooms, isolation rooms, patient care areas and laboratories. When used as directed, it will also effectively clean and decontaminate critical and semi-critical instrumentation. CaviCide is safe for use on non-porous surfaces. It is perfect for cleaning and decontamination of environmental and medical device surfaces.
Catalog No. 50-209-1723
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.