Learn More
Essendant EARPLUGSCLSCUNCLRD
Supplier:  Essendant MMM3101101
Encompass_Preferred
The world s first foam earplugs and are still one of the most widely used earplugs today Classic Plus earplugs are slightly longer than regular E-A-R Classic earplugs to fit deeper into the ear canal and provide higher noise reduction Slow recovery low pressure foam fits snugly to create a noise barrier that helps protect hearing from loud sounds The earplugs are both flame resistant and moisture resistant Bright yellow color allows supervisors to conduct a quick visual hearing protection compliance check Recommended applications include chipping chiseling drilling furnace operations grinding heavy equipment operations machining pouring/casting power fastening riveting sanding sawing and welding and more Test compatible with the 3M E-A-Rfit Validation System CSA Class AL Research suggests that many users will receive less noise reduction than indicated by the NRR due to variation in earplug fit earplug fitting skill and motivation of the user It is recommend
Catalog No. 50-000-33085
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.