Learn More
Essendant Speedball Heavy-Duty Cleaner, Citrus, Liquid, 1qt. Spray Bottle, 12/CT
Supplier:  Essendant DVO95891164
Encompass_Preferred
Non-butyl, heavy-duty product cleans the toughest soils. Non-acid formula is recommended for use on most washable surfaces. Convenient spray-and-wipe cleaning. Not for use on water-based paint, aluminum or glass.
Catalog No. 27-905-55
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.