Learn More
Essendant FogShield Disposable Lens Cleaning Station, 12 oz Bottle, 1,425 Tissues/Box
Supplier:  Essendant BAL8577
Encompass_Preferred
FogShield protection lens cleaning kit provides an excellent option if your facility has extreme variations in temperatures or harsh conditions. Easy-to-use FogShield is applied in two steps ¿ first spray and clean the surface with FogShield XP, then use a dry tissue to buff, polish, and coat the surface for extreme protection. Combining the effective cleaning of all glass and plastic lenses with extreme anti-fog protection, the FogShield XP Lens Cleaning Station can keep employees vision clear in all conditions. The durable corrugated station is moisture resistant and can be hung on a wall or displayed on a counter. High capacity station with 12 oz. spray bottle with silicone and 1,425 lint free absorbent tissues is great for high traffic areas.
Catalog No. 27-937-15
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.