Learn More
Essendant Finished Floor Cleaner, Lemon Scent, 1 gal Bottle, 3/Carton
Supplier:  Essendant PGC02621CT
Encompass_Preferred
Mr. Clean Finished Floor Cleaner is a powerful professional floor cleaner liquid that removes tough ground-in dirt and preserves shine. Mr. Clean multi surface cleaner can be used on ceramic and vinyl tile, terrazzo flooring, and varnished hardwoods. This versatile floor cleaning product makes cleaning easy because it penetrates and dissolves tough grease and dirt on contact and is designed to extend life between scrub and recoat. Recommended dilution: manual (1:128), automatic machines (1:512).
Catalog No. 26-792-47
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.