Learn More
Essendant Quick Clean Griddle Liquid, 3.2 oz Packet, 40/Carton
Supplier:  Essendant MMM29603
Encompass_Preferred
Effectively removes baked-on food and cooking oil from griddles. Convenient one-step cleaning requires no rinsing, saving you time and labor. Designed for use on medium-hot griddles: 300? to 350? F. Caustic soda- and potash-free.
Catalog No. 26-797-36
Only null left
Add to Cart
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Your feedback has been submitted: Thank you for helping us improve our website.