Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Cytiva CleanGel for IEF

Catalog No. 45000442 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
5
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
45-000-442 5
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 45-000-442 Supplier Cytiva Supplier No. 18103532

missing translation for 'validMainframeMsg'

Perform sensitive procedures using precast gels that are carefully washed and dried after casting to remove catalysts and unpolymerized material

  • Customize IEF separations by reswelling in a suitable mixture of carrier ampholytes and other additives such as detergents or denaturants
  • Handle mechanically stable, plastic-backed gels with confidence
  • Offer improved gel performance with faster and more stable gradients, fewer artifacts and easier zymogram assays
  • Before use, rehydrate using the convenient GelPool accessory (Mfr. No. 18-1031-58); swelling returns the gel to its original thickness of 0.5mm
  • Gel composition: Polyacrylamide gel (5%T/3%C)

TRUSTED_SUSTAINABILITY

Specifications

Description CleanGel IEF
Color Clear
Format Precast Gel
Separation Type Isoelectric Focusing
For Use With (Application) Analytical chemistry, Research CleanGel IEF gels offer improved gel performance with faster and more stable gradients, fewer artifacts and easier zymogram assays
Material Polyester film
Quantity 5
Storage Requirements -20°C
Thickness (English) 0.019 in.
Thickness (Metric) 0.05 cm
Length (Metric) 110 mm
Product Type Precast Gel
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.