Learn More
New Pig CleanMax™ Cleanroom Coverall
SDP
Catalog No. 17987902 Shop All New Pig Corporation Products
Change view
Click to view available options
Size:
Medium
Large
X-Large
2X-Large
3X-Large
4X-Large
5X-Large
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-987-902 4X-Large
17-987-903 5X-Large
17-987-898 Large
17-987-897 Medium
17-987-899 X-Large
17-987-900 2X-Large
17-987-901 3X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. 17-987-902 Supplier New Pig Supplier No. WPL30034XL
Only null left
Add to Cart
Add to Cart
Help protect yourself in cleanrooms from certain pathogens and viruses by wearing this bound-seam coverall with attached hood and boots
  • The coverall is constructed from a composite fabric of high-density polyethylene film laminated to a continuous filament polypropylene nonwoven.
  • This provides a strong barrier against particles, microorganisms, and liquid splashes.
  • The material is designed to be low-linting and to shed minimal particles, which is essential for maintaining the integrity of ISO Class 5-8 cleanroom environments.
  • Despite its strong barrier properties, the fabric is engineered to be breathable, allowing for the passage of moisture vapor and helping to reduce heat stress for the wearer.
  • Includes features such as a front zipper with a storm flap, elastic wrists, and elastic ankles to provide a secure fit and prevent the ingress of contaminants.
  • The CleanMax™ product line is often positioned as an economical alternative to more expensive cleanroom garments, providing a high level of protection at a lower cost for applications that require a balance of performance and value.
Specifications
Size 4X-Large