Learn More
Uniform Technology™ Cleanroom Boot Altessa Grid ISO 5 Cleanroom Boots
Catalog No. 17779038 Shop All Protective Industrial Products Inc. Products
Change view
Click to view available options
Size:
X-Small
Small
Medium
X-Large
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
17-779-038 X-Small
17-779-039 Small
17-779-040 Medium
17-779-041 -
17-779-042 X-Large
17-779-043 -
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
This item is not returnable. View return policy
Catalog No. 17-779-038 Supplier Uniform Technology™ Supplier No. CBPX74WHXS
Add to Cart
Add to Cart
This item is not returnable. View return policy
This cleanroom boot cover is ideal for contamination prevention in general, high-tech, semiconductor, optical manufacturing, and inspection applications.
  • Altessa Grid Fabric is made of 99% polyester to 1% carbon blend
  • Color-coded webbing across toe with strap
  • Webbing around ankle with side release buckle closure
  • 1 in. elastic and webbing top closure with snap adjustment
  • X-Stat ESD sole with twill reinforcement
  • Upper is sewn on with conductive thread
  • Meets standards required for use in ISO 5 (Class 100) to ISO 8 (Class 100,000) cleanrooms and other controlled environments
Specifications
Color White
Material Polyblend
Size X-Small
Waterproof Water Repellent
Product Type Cleanroom Boots
Cleanroom Class ISO 5 (Class 100)