Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Keystone Cleanroom Connection Bouffant Cap
SDP

Catalog No. 18366657 Shop All New Pig Corporation Products
Change view
Click to view available options
Size:
24 in.
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-366-657 24 in.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 18-366-657 Supplier Keystone Supplier No. WPL892
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Bouffant cap meant to prevent hair from falling onto cleanroom worksurfaces, as well as to protect your hair from contaminants.

  • Latex- and lint-free
  • Protect your work area and your hair
  • Ideal for cleanrooms as well as chemotherapy-related tasks
  • Class 1000 ISO6+
  • Breathable and soft for comfort over longer periods
  • Non-sterile
  • 100 per bag

Specifications

Product Type Cleanroom Bouffant
Size 24 in.
Material Polypropylene (Non-woven)
Color White
Disposable Single-use
Latex Free Yes
Sterility Non-sterile
Quantity 100/Cs.
Cleanroom Class Class 1000 (ISO 6)
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.