Learn More
Biomedix Cleanroom Connection Chemotherapy Gown
SDP
Catalog No. 18366663 Shop All New Pig Corporation Products
Change view
Click to view available options
Size:
Small
Large
X-Large
3X-Large
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Size
18-366-663 3X-Large
18-366-661 Small
18-366-660 Large
18-366-662 X-Large
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. 18-366-663 Supplier Biomedix Supplier No. WPL981XXXL
Add to Cart
Add to Cart
Sterile gown for safely handling chemotherapy and other cytotoxic drugs.
  • Sterile gowns designed to protect you while handling chemotherapy and other cytotoxic drugs
  • Ideal for oncology centers, hospitals, pharmacies and more
  • Taped seams and closed-front help make these polypropylene-coated gowns impervious
  • Elastic wrists and tie-back closure add protection
  • Comfortable, breathable fabric that is lint- and latex-free
  • Sterile cleanroom garments are designed to protect a cleanroom environment from contamination by viable organisms
  • ISO 5 Class 100 compliant
  • USP General Chapter 800 compliant
  • Tested by the ASTM D6978-05 method for chemotherapy permeation
Specifications
No. of Pockets None
Gender Unisex
Size 3X-Large
Product Type Controlled Environments Gown
Material Polypropylene
Color White
Closure Type Back Ties
Disposable Single-use
Sterility Sterile
Cuff Style Elastic
Latex Free Yes
Sleeve Style Long Sleeve
Autoclavable Not Autoclavable